Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Exosc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163570.1 | NP_001157042.1 | 7 | 726 | ||
NM_027148.3 | NP_081424.3 | 7 | 714 | |||
GenBank mRNA | AJ505005.1 | - | 7 | 374 | ||
AK009584.1 | - | 7 | 386 | |||
AK047257.1 | - | 5 | 1284 | |||
AK169719.1 | - | 5 | 317 | |||
AK220227.1 | - | 7 | 714 | |||
BC004769.1 | - | 5 | 954 | |||
BC059089.1 | - | 7 | 743 | 6509820 | ||
BC067250.1 | - | 6 | 357 |