Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAST2 SIRNA Assays ›
Gene Symbol:
MAST2
Gene Name:
microtubule associated serine/threonine kinase 2
Gene Aliases:
MAST205, MTSSK
Chromosome Location:
Chr.1: 45786992 - 46036124 on Build GRCh38
UniGene ID:
Hs.319481
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
103517
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAST2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001319245.1
NP_001306174.1
12
1648
NM_001324320.1
NP_001311249.1
13
1675
NM_001324321.1
NP_001311250.1
12
1761
NM_015112.2
NP_055927.2
12
1648
XM_005270655.3
XP_005270712.1
11
1368
XM_005270656.4
XP_005270713.1
11
1237
XM_006710477.3
XP_006710540.1
10
1045
XM_011541059.2
XP_011539361.1
14
1882
XM_011541061.2
XP_011539363.1
14
1882
XM_011541062.2
XP_011539364.1
13
1861
XM_011541063.1
XP_011539365.1
14
1446
XM_011541064.2
XP_011539366.1
14
1443
XM_011541065.1
XP_011539367.1
14
1545
XM_011541067.2
XP_011539369.1
10
1359
XM_011541068.1
XP_011539370.1
9
976
XM_011541069.2
XP_011539371.1
14
1882
XM_017000752.1
XP_016856241.1
12
1875
XM_017000753.1
XP_016856242.1
11
1209
XM_017000754.1
XP_016856243.1
10
1188
XM_017000755.1
XP_016856244.1
11
1680
GenBank mRNA
AB047005.1
-
12
1648
AK027123.1
-
10
1187
AK096519.1
-
11
972
AK309510.1
-
11
1370
BC006166.1
-
4
316
BC015816.2
-
4
316
BC065499.1
-
12
1622
5499830
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
cytoskeleton organization
peptidyl-serine phosphorylation
intracellular signal transduction
regulation of interleukin-12 biosynthetic process
spermatid differentiation
magnesium ion binding
protein serine/threonine kinase activity
protein binding
ATP binding
microtubule binding
phosphatase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-4b6rj:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0