Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MMP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002427.3 | NP_002418.1 | 9 | 1313 | ||
GenBank mRNA | AK222748.1 | - | 9 | 1313 | ||
AK292211.1 | - | 9 | 1313 | |||
AK315341.1 | - | 9 | 1311 | |||
AY256443.1 | - | 9 | 1284 | |||
BC067522.1 | - | 9 | 1292 | 7002184 | ||
BC074807.2 | - | 9 | 1303 | 30915315 | ||
BC074808.2 | - | 9 | 1303 | 30915529 | ||
X75308.1 | - | 9 | 1288 | |||
X81334.1 | - | 9 | 1311 |