Search
Search

CPB2| Gene Symbol: CPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278541.1 | NP_001265470.1 | 1, 2 | 147 | ||
| NM_001872.4 | NP_001863.3 | 1, 2 | 147 | |||
| GenBank mRNA | AK290829.1 | - | 1, 2 | 95 | ||
| AU100156.1 | - | 1, 2 | 147 | |||
| BC007057.1 | - | 1, 2 | 104 | 3934520 | ||
| BT006936.1 | - | 1, 2 | 80 | |||
| M75106.1 | - | 1, 2 | 99 | |||