Search
Search

FADS1| Gene Symbol: FADS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013402.4 | NP_037534.3 | 12 | 2431 | ||
| XM_011545022.2 | XP_011543324.1 | 12 | 2105 | |||
| GenBank mRNA | AF035284.1 | - | 1 | 377 | ||
| AF084558.1 | - | 12 | 2216 | |||
| AL512760.1 | - | 12 | 2212 | |||
| AL834479.1 | - | 4 | 1075 | |||