Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SLC9B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300754.1 | NP_001287683.1 | 7, 8 | 1099 | ||
NM_001300756.1 | NP_001287685.1 | 7, 8 | 1465 | |||
NM_178833.5 | NP_849155.2 | 8, 9 | 1636 | |||
XM_006714085.2 | XP_006714148.1 | 8, 9 | 1635 | |||
XM_006714086.3 | XP_006714149.1 | 7, 8 | 1464 | |||
GenBank mRNA | AK172823.1 | - | 8, 9 | 1636 | ||
AK297010.1 | - | 7, 8 | 1010 | |||
AK299888.1 | - | 7, 8 | 1024 | |||
BC009732.1 | - | 5, 6 | 704 | |||
BC047447.1 | - | 8, 9 | 1166 | 5295722 |