Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ZAP70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079.3 | NP_001070.2 | 14 | 2333 | ||
NM_207519.1 | NP_997402.1 | 6 | 1351 | |||
XM_017004867.1 | XP_016860356.1 | 16 | 3734 | |||
XM_017004868.1 | XP_016860357.1 | 16 | 3716 | |||
GenBank mRNA | AL832287.1 | - | 7 | 3178 | ||
BC039039.1 | - | 13 | 1987 | 5168073 | ||
BC053878.1 | - | 14 | 2309 | 5757161 | ||
L05148.1 | - | 14 | 2335 |