Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PXK SIRNA Assays ›
Gene Symbol:
PXK
Gene Name:
PX domain containing serine/threonine kinase like
Gene Aliases:
MONaKA
Chromosome Location:
Chr.3: 58332868 - 58426127 on Build GRCh38
UniGene ID:
Hs.190544
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
110964
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PXK
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289095.1
NP_001276024.1
5
575
NM_001289096.1
NP_001276025.1
4
527
NM_001289098.1
NP_001276027.1
6
626
NM_001289099.1
NP_001276028.1
6
731
NM_001289100.1
NP_001276029.1
4
527
NM_001289101.1
NP_001276030.1
3
340
NM_017771.4
NP_060241.2
6
626
XM_005265250.4
XP_005265307.1
6
626
XM_005265252.4
XP_005265309.1
6
626
XM_005265255.3
XP_005265312.1
6
626
XM_005265256.3
XP_005265313.1
6
626
XM_017006671.1
XP_016862160.1
5
517
XM_017006672.1
XP_016862161.1
5
517
XM_017006673.1
XP_016862162.1
4
518
XM_017006674.1
XP_016862163.1
5
517
XM_017006675.1
XP_016862164.1
6
626
XM_017006676.1
XP_016862165.1
6
626
XM_017006677.1
XP_016862166.1
6
626
XM_017006678.1
XP_016862167.1
6
626
XM_017006679.1
XP_016862168.1
6
626
XM_017006680.1
XP_016862169.1
6
626
XM_017006681.1
XP_016862170.1
5
517
XM_017006682.1
XP_016862171.1
5
517
XM_017006683.1
XP_016862172.1
5
517
XM_017006684.1
XP_016862173.1
5
517
XM_017006685.1
XP_016862174.1
5
517
XM_017006686.1
XP_016862175.1
7
740
XM_017006687.1
XP_016862176.1
7
738
XM_017006688.1
XP_016862177.1
4
502
XM_017006689.1
XP_016862178.1
4
492
XM_017006690.1
XP_016862179.1
4
483
XM_017006691.1
XP_016862180.1
3
339
XM_017006692.1
XP_016862181.1
7
777
XM_017006693.1
XP_016862182.1
7
777
XM_017006694.1
XP_016862183.1
7
671
XM_017006695.1
XP_016862184.1
7
777
XM_017006696.1
XP_016862185.1
3
339
XM_017006697.1
XP_016862186.1
3
334
XM_017006698.1
XP_016862187.1
3
332
XM_017006699.1
XP_016862188.1
3
344
XM_017006700.1
XP_016862189.1
3
362
XM_017006701.1
XP_016862190.1
3
339
XM_017006702.1
XP_016862191.1
3
339
XM_017006703.1
XP_016862192.1
3
339
XM_017006704.1
XP_016862193.1
3
339
GenBank mRNA
AB209688.1
-
6
604
AF399753.1
-
6
527
AK000342.1
-
6
624
AK131385.1
-
5
517
AK295542.1
-
4
519
AK296034.1
-
3
198
AK296215.1
-
3
275
AY274811.1
-
6
626
AY437879.1
-
4
470
AY847220.1
-
6
634
AY847221.1
-
3
243
BC008943.2
-
6
607
BC014479.1
-
6
542
4856139
DQ124707.1
-
6
527
DQ124708.1
-
6
527
KF774202.1
-
6
636
KF774203.1
-
6
636
KF774204.1
-
6
711
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
inflammatory response
negative regulation of ATPase activity
regulation of membrane potential
negative regulation of ion transport
modulation of synaptic transmission
nucleotide binding
actin binding
protein kinase activity
ATP binding
protein C-terminus binding
phosphatidylinositol binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0