Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TNC SIRNA Assays ›
          Gene Symbol:
          TNC
          Gene Name:
          tenascin C
          Gene Aliases:
          150-225, DFNA56, GMEM, GP, HXB, JI, TN, TN-C
          Chromosome Location:
          Chr.9: 115019575 - 115118257 on Build GRCh38
          UniGene ID:
          Hs.143250
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 11165

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TNC
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002160.3 NP_002151.2 3 1773
          XM_005251972.3 XP_005252029.1 3 1723
          XM_005251973.3 XP_005252030.1 3 1723
          XM_005251974.3 XP_005252031.1 3 1723
          XM_005251975.3 XP_005252032.1 3 1723
          XM_006717096.3 XP_006717159.1 3 1723
          XM_006717097.3 XP_006717160.1 3 1723
          XM_006717098.3 XP_006717161.1 3 1723
          XM_006717101.3 XP_006717164.1 3 1723
          XM_011518625.2 XP_011516927.1 3 1723
          XM_011518626.2 XP_011516928.1 3 1723
          XM_011518628.2 XP_011516930.1 3 1723
          XM_011518629.2 XP_011516931.1 3 1723
          XM_017014678.1 XP_016870167.1 3 1723
          XM_017014679.1 XP_016870168.1 3 1723
          XM_017014680.1 XP_016870169.1 3 1723
          XM_017014681.1 XP_016870170.1 3 1723
          GenBank mRNA AB210038.1 - 3 1649
          AK304015.1 - 3 1669
          BC151843.1 - 3 1641 8860362
          GU473235.1 - 2 1361
          M24630.1 - 1 71
          M55618.1 - 2 1496
          X56160.1 - 2 1412
          X78565.1 - 3 1674

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          extracellular matrix protein

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          cell adhesion
          negative regulation of cell adhesion
          neuromuscular junction development
          positive regulation of cell proliferation
          response to wounding
          response to mechanical stimulus
          positive regulation of gene expression
          peripheral nervous system axon regeneration
          extracellular matrix organization
          wound healing
          odontogenesis of dentin-containing tooth
          response to ethanol
          bud outgrowth involved in lung branching
          mesenchymal-epithelial cell signaling involved in prostate gland development
          prostate gland epithelium morphogenesis
          cellular response to retinoic acid
          cellular response to vitamin D
          response to fibroblast growth factor
          cellular response to prostaglandin D stimulus
          syndecan binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8djhq:80/100.66.76.44:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0