Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CHRNA7 SIRNA Assays ›
          Gene Symbol:
          CHRNA7
          Gene Name:
          cholinergic receptor nicotinic alpha 7 subunit
          Gene Aliases:
          CHRNA7-2, NACHRA7
          Chromosome Location:
          Chr.15: 32030462 - 32172521 on Build GRCh38
          UniGene ID:
          Hs.511772
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 114074

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CHRNA7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000746.5 NP_000737.1 2 268
          NM_001190455.2 NP_001177384.1 2 355
          NR_046324.1 2 268
          XM_011521178.2 XP_011519480.1 2 268
          XM_017021882.1 XP_016877371.1 2 206
          XM_017021883.1 XP_016877372.1 2 206
          XM_017021884.1 XP_016877373.1 2 289
          GenBank mRNA AF385585.1 - 2 170
          AJ586911.1 - 2 173
          AK292069.1 - 2 238
          AK294229.1 - 2 350
          AY909581.1 - 1 138
          BC037571.1 - 2 195 5245812
          DA583811.1 - 2 268
          EF405802.1 - 2 206
          EF405803.1 - 1 22
          EF405804.1 - 1 15
          EF405812.1 - 2 174
          EF405813.1 - 2 174
          EF405814.1 - 2 206
          EF405815.1 - 1 22
          EF405819.1 - 1 13
          EF405820.1 - 1 13
          EF405821.1 - 1 13
          L25827.1 - 1 133
          U40583.1 - 2 163
          U62436.1 - 2 228
          X70297.1 - 2 259
          Y08420.1 - 2 156
          Z23141.1 - 1 87

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ligand-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          response to hypoxia
          ion transport
          calcium ion transport
          cellular calcium ion homeostasis
          signal transduction
          synaptic transmission, cholinergic
          memory
          positive regulation of cell proliferation
          negative regulation of tumor necrosis factor production
          ion transmembrane transport
          response to nicotine
          positive regulation of angiogenesis
          cognition
          cation transmembrane transport
          beta-amyloid binding
          acetylcholine-activated cation-selective channel activity
          ligand-gated ion channel activity
          acetylcholine receptor activity
          toxic substance binding
          chloride channel regulator activity
          acetylcholine-gated cation channel activity
          acetylcholine binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-wf5dr:80/100.66.77.5:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0