Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ENPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006208.2 | NP_006199.2 | 20 | 1987 | ||
XM_011535896.1 | XP_011534198.1 | 11 | 916 | |||
GenBank mRNA | AK314870.1 | - | 20 | 1814 | ||
BC059375.1 | - | 20 | 1987 | 30341060 | ||
D12485.1 | - | 20 | 1983 | |||
M57736.1 | - | 20 | 1974 |