Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MYO7A SIRNA Assays ›
          Gene Symbol:
          MYO7A
          Gene Name:
          myosin VIIA
          Gene Aliases:
          DFNA11, DFNB2, MYOVIIA, MYU7A, NSRD2, USH1B
          Chromosome Location:
          Chr.11: 77128192 - 77215241 on Build GRCh38
          UniGene ID:
          Hs.370421
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 118350

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MYO7A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000260.3 NP_000251.3 13 1722
          NM_001127179.2 NP_001120651.2 13 1722
          NM_001127180.1 NP_001120652.1 13 1716
          XM_011545044.2 XP_011543346.1 13 1794
          XM_011545046.2 XP_011543348.2 12 1555
          XM_011545050.2 XP_011543352.1 10 1409
          XM_017017778.1 XP_016873267.1 12 1555
          XM_017017779.1 XP_016873268.1 12 1555
          XM_017017780.1 XP_016873269.1 12 1555
          XM_017017781.1 XP_016873270.1 12 1555
          XM_017017782.1 XP_016873271.1 12 1555
          XM_017017783.1 XP_016873272.1 12 1555
          XM_017017784.1 XP_016873273.1 12 1555
          XM_017017785.1 XP_016873274.1 10 1324
          XM_017017786.1 XP_016873275.1 12 1555
          XM_017017787.1 XP_016873276.1 12 1555
          XM_017017788.1 XP_016873277.1 12 1555
          GenBank mRNA BC144519.1 - 14 1764
          U34227.1 - 13 1717
          U39226.1 - 13 1722
          U55208.1 - 13 1717
          U55209.1 - 13 1717

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_153473, XM_006229743, XM_006229744, XM_006229742, AB091825.

          Panther Classification:

          Molecular Function -

          actin binding motor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          phagolysosome assembly
          intracellular protein transport
          lysosome organization
          visual perception
          sensory perception of sound
          actin filament-based movement
          eye photoreceptor cell development
          post-embryonic organ morphogenesis
          sensory perception of light stimulus
          equilibrioception
          pigment granule transport
          auditory receptor cell stereocilium organization
          microfilament motor activity
          protein binding
          calmodulin binding
          ATP binding
          protein domain specific binding
          spectrin binding
          actin-dependent ATPase activity
          protein homodimerization activity
          ADP binding
          protein N-terminus binding
          actin filament binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0