Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SFRP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003014.3 | NP_003005.2 | 6 | 1436 | ||
GenBank mRNA | AF026692.1 | - | 6 | 1307 | ||
AK302357.1 | - | 6 | 1358 | |||
AK303977.1 | - | 7 | 1256 | |||
BC047684.1 | - | 6 | 1294 | 5742936 | ||
BC058911.1 | - | 6 | 1314 | 5228231 | ||
DQ420628.1 | - | 6 | 1069 |
Set Membership: |
G-protein coupled receptor |