Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MAPKAPK5 SIRNA Assays ›
          Gene Symbol:
          MAPKAPK5
          Gene Name:
          mitogen-activated protein kinase-activated protein kinase 5
          Gene Aliases:
          MAPKAP-K5, MK-5, MK5, PRAK
          Chromosome Location:
          Chr.12: 111842228 - 111902225 on Build GRCh38
          UniGene ID:
          Hs.333120
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 1229

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price 417.00
          Your Price
          Online offer:
          417.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MAPKAPK5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003668.3 NP_003659.2 3, 4 700
          NM_139078.2 NP_620777.1 3, 4 700
          XM_017020125.1 XP_016875614.1 3, 4 192
          XM_017020132.1 XP_016875621.1 3, 4 558
          XM_017020133.1 XP_016875622.1 3, 4 454
          XM_017020134.1 XP_016875623.1 3, 4 280
          XM_017020135.1 XP_016875624.1 3, 4 453
          XM_017020136.1 XP_016875625.1 3, 4 454
          XM_017020137.1 XP_016875626.1 3, 4 453
          XM_017020139.1 XP_016875628.1 3, 4 453
          GenBank mRNA AK122767.1 - 3, 4 950
          AW780199.1 - 3, 4 454
          BC000833.1 - 2, 3 143
          BC041049.1 - 3, 4 687 4909021
          BC047284.1 - 3, 4 368 4997201

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          activation of MAPK activity
          regulation of translation
          signal transduction
          Ras protein signal transduction
          peptidyl-serine phosphorylation
          negative regulation of TOR signaling
          positive regulation of telomere maintenance via telomerase
          protein autophosphorylation
          positive regulation of telomerase activity
          stress-induced premature senescence
          regulation of signal transduction by p53 class mediator
          positive regulation of telomere capping
          p53 binding
          protein serine/threonine kinase activity
          calmodulin-dependent protein kinase activity
          MAP kinase kinase activity
          protein binding
          calmodulin binding
          ATP binding
          calcium-dependent protein serine/threonine kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0