Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TMCO4 SIRNA Assays ›
          Gene Symbol:
          TMCO4
          Gene Name:
          transmembrane and coiled-coil domains 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 19682213 - 19800385 on Build GRCh38
          UniGene ID:
          Hs.656313
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 124353

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TMCO4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_181719.4 NP_859070.3 4, 5 419
          XM_005245822.4 XP_005245879.1 4, 5 435
          XM_005245824.4 XP_005245881.1 4, 5 499
          XM_005245825.4 XP_005245882.1 3, 4 300
          XM_005245827.3 XP_005245884.1 4, 5 435
          XM_006710551.3 XP_006710614.1 4, 5 524
          XM_011541179.2 XP_011539481.1 7, 8 894
          XM_011541180.2 XP_011539482.1 7, 8 1049
          XM_011541181.1 XP_011539483.1 6, 7 723
          XM_011541185.2 XP_011539487.1 4, 5 442
          XM_011541186.2 XP_011539488.1 4, 5 539
          XM_017000913.1 XP_016856402.1 4, 5 443
          XM_017000914.1 XP_016856403.1 6, 7 943
          XM_017000915.1 XP_016856404.1 6, 7 665
          XM_017000916.1 XP_016856405.1 4, 5 577
          XM_017000917.1 XP_016856406.1 5, 6 792
          XM_017000918.1 XP_016856407.1 6, 7 989
          XM_017000919.1 XP_016856408.1 5, 6 716
          XM_017000920.1 XP_016856409.1 4, 5 934
          XM_017000921.1 XP_016856410.1 5, 6 741
          GenBank mRNA AK309310.1 - 4, 5 435
          AK309900.1 - 2, 3 234
          AL049846.1 - 1, 2 185
          AL049847.1 - 1, 2 183
          AX775873.1 - 3, 4 345
          AX775875.1 - 3, 4 448
          AX775877.1 - 3, 4 448
          BC053600.1 - 2, 3 278 5744211
          BX640987.1 - 4, 5 420

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-7xtrm:80/100.66.78.85:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0