Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: WDSUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128212.2 | NP_001121684.1 | 11 | 1857 | ||
NM_001128213.1 | NP_001121685.1 | 11 | 1873 | |||
NM_001307994.1 | NP_001294923.1 | 7 | 1581 | |||
NM_152528.2 | NP_689741.2 | 11 | 1765 | |||
XM_005246355.2 | XP_005246412.2 | 10 | 1710 | |||
XM_006712341.3 | XP_006712404.1 | 10 | 1345 | |||
XM_011510753.2 | XP_011509055.1 | 11 | 1817 | |||
XM_011510754.2 | XP_011509056.1 | 11 | 1551 | |||
XM_017003482.1 | XP_016858971.1 | 6 | 1482 | |||
XM_017003483.1 | XP_016858972.1 | 10 | 1672 | |||
GenBank mRNA | AK093494.1 | - | 11 | 1764 | ||
AK129983.1 | - | 11 | 1872 | |||
BC029520.1 | - | 7 | 1485 | 5301559 |