Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EZR SIRNA Assays ›
          Gene Symbol:
          EZR
          Gene Name:
          ezrin
          Gene Aliases:
          CVIL, CVL, HEL-S-105, VIL2
          Chromosome Location:
          Chr.6: 158765741 - 158819424 on Build GRCh38
          UniGene ID:
          Hs.487027
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 13110

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: EZR
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001111077.1 NP_001104547.1 4 349
          NM_003379.4 NP_003370.2 3 383
          GenBank mRNA AK296738.1 - 4 312
          AK312597.1 - 4 293
          AL162086.1 - 4 301
          BC013903.2 - 3 300 2959399
          BC068458.1 - 3 309 4819793
          DA754543.1 - 4 349
          FJ224318.1 - 3 285
          J05021.1 - 3 244
          X51521.1 - 3 285

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_019357, AF450298, BC081958, AY428869.
          This siRNA also targets the following transcripts in Mouse: NM_009510, BC048181, AK168410, X60671, AK135353, and others.

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          intestinal D-glucose absorption
          sphingosine-1-phosphate signaling pathway
          cytoskeletal anchoring at plasma membrane
          leukocyte cell-cell adhesion
          axon guidance
          regulation of cell shape
          regulation of cell size
          positive regulation of gene expression
          protein kinase A signaling
          gland morphogenesis
          membrane to membrane docking
          microvillus assembly
          astral microtubule organization
          actin cytoskeleton reorganization
          receptor internalization
          regulation of microvillus length
          regulation of actin cytoskeleton organization
          cellular protein complex localization
          positive regulation of multicellular organism growth
          cortical microtubule organization
          establishment of epithelial cell apical/basal polarity
          filopodium assembly
          phosphatidylinositol-mediated signaling
          positive regulation of protein secretion
          negative regulation of T cell receptor signaling pathway
          actin filament bundle assembly
          establishment of centrosome localization
          establishment of endothelial barrier
          negative regulation of ERK1 and ERK2 cascade
          cellular response to cAMP
          protein localization to cell cortex
          establishment of protein localization to plasma membrane
          positive regulation of establishment of protein localization to plasma membrane
          negative regulation of interleukin-2 secretion
          regulation of NIK/NF-kappaB signaling
          regulation of organelle assembly
          terminal web assembly
          positive regulation of protein localization to early endosome
          positive regulation of cellular protein catabolic process
          negative regulation of p38MAPK cascade
          positive regulation of early endosome to late endosome transport
          actin binding
          protein binding
          microtubule binding
          protein domain specific binding
          protein kinase A catalytic subunit binding
          protein kinase A regulatory subunit binding
          S100 protein binding
          poly(A) RNA binding
          cell adhesion molecule binding
          actin filament binding
          ATPase binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-sskjb:80/100.66.75.127:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0