Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RSPH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031924.4 | NP_114130.3 | 4 | 1048 | ||
XM_017011347.1 | XP_016866836.1 | 3 | 369 | |||
GenBank mRNA | AF353618.1 | - | 4 | 647 | ||
AK057931.1 | - | 4 | 1069 | |||
BC050604.1 | - | 4 | 1048 | 6161669 |