Search
Search

STMN2| Gene Symbol: STMN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199214.1 | NP_001186143.1 | 3 | 519 | ||
| NM_007029.3 | NP_008960.2 | 3 | 519 | |||
| XM_005251142.2 | XP_005251199.1 | 4 | 143 | |||
| GenBank mRNA | AK292737.1 | - | 3 | 191 | ||
| AK297485.1 | - | 3 | 200 | |||
| AK297594.1 | - | 3 | 213 | |||
| AK299500.1 | - | 3 | 228 | |||
| AK308557.1 | - | 3 | 202 | |||
| AK309130.1 | - | 3 | 406 | |||
| BC006302.2 | - | 3 | 188 | 4100671 | ||
| BQ069488.1 | - | 3 | 181 | |||
| BT020034.1 | - | 2 | 137 | |||
| CR456833.1 | - | 2 | 137 | |||
| D50375.1 | - | 3 | 254 | |||
| DC319376.1 | - | 3 | 519 | |||
| S82024.1 | - | 3 | 165 | |||