Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYNE1 SIRNA Assays ›
Gene Symbol:
SYNE1
Gene Name:
spectrin repeat containing nuclear envelope protein 1
Gene Aliases:
8B, ARCA1, C6orf98, CPG2, EDMD4, MYNE1, Nesp1, SCAR8, dJ45H2.2
Chromosome Location:
Chr.6: 152121684 - 152637399 on Build GRCh38
UniGene ID:
Hs.12967
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
137092
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
286.00
Your Price
Online offer:
277.65
277.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SYNE1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_033071.3
NP_149062.1
129
23756
NM_182961.3
NP_892006.3
130
24134
XM_006715407.1
XP_006715470.1
130
24047
XM_006715408.2
XP_006715471.1
130
24039
XM_006715409.1
XP_006715472.1
129
24026
XM_006715410.2
XP_006715473.1
130
24051
XM_006715411.1
XP_006715474.1
129
23996
XM_006715412.2
XP_006715475.1
130
24036
XM_006715413.2
XP_006715476.1
130
24051
XM_006715414.1
XP_006715477.1
128
23975
XM_006715415.2
XP_006715478.1
130
24051
XM_006715416.2
XP_006715479.1
130
24036
XM_006715417.2
XP_006715480.1
129
23910
XM_006715420.2
XP_006715483.1
129
23898
XM_006715421.2
XP_006715484.1
129
23895
XM_006715422.1
XP_006715485.1
129
23888
XM_006715423.2
XP_006715486.1
130
24051
XM_006715424.2
XP_006715487.1
130
24051
XM_006715425.2
XP_006715488.1
130
24051
XM_011535641.2
XP_011533943.1
130
24048
XM_011535642.2
XP_011533944.1
130
24036
XM_011535643.1
XP_011533945.1
129
23882
XM_011535644.1
XP_011533946.1
116
22098
XM_011535645.2
XP_011533947.1
112
21491
XM_017010608.1
XP_016866097.1
131
24166
XM_017010609.1
XP_016866098.1
130
23973
XM_017010610.1
XP_016866099.1
129
23952
XM_017010611.1
XP_016866100.1
129
23755
XM_017010612.1
XP_016866101.1
128
23679
XM_017010613.1
XP_016866102.1
130
24048
XM_017010614.1
XP_016866103.1
129
23895
XM_017010615.1
XP_016866104.1
129
23895
XM_017010616.1
XP_016866105.1
130
24051
XM_017010617.1
XP_016866106.1
130
24048
XM_017010618.1
XP_016866107.1
130
24036
XM_017010619.1
XP_016866108.1
116
22130
GenBank mRNA
AB018339.2
-
18
2771
AF444779.1
-
5
644
AF495910.1
-
130
24134
AF535142.1
-
129
23756
AK122691.1
-
10
1414
AK127076.1
-
14
2111
AK304825.1
-
3
336
AK316101.1
-
3
351
AY061755.1
-
45
7224
AY061756.1
-
2
214
AY184206.1
-
43
7119
BC090927.1
-
3
396
BX537837.1
-
12
1712
BX647837.1
-
15
2306
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
nucleus organization
Golgi organization
establishment of nucleus localization
muscle cell differentiation
cytoskeletal anchoring at nuclear membrane
nuclear matrix anchoring at nuclear membrane
actin binding
protein binding
lamin binding
protein homodimerization activity
poly(A) RNA binding
actin filament binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0