Search
Search

CD101| Gene Symbol: CD101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256106.2 | NP_001243035.1 | 3 | 557 | ||
| NM_001256109.2 | NP_001243038.1 | 3 | 557 | |||
| NM_004258.5 | NP_004249.2 | 3 | 557 | |||
| GenBank mRNA | AK307542.1 | - | 3 | 557 | ||
| BC130327.1 | - | 3 | 518 | 40146358 | ||
| BC136484.1 | - | 3 | 518 | 9020473 | ||
| BC171768.1 | - | 3 | 518 | 9054422 | ||
| DA955029.1 | - | 3 | 548 | |||
| Z33642.1 | - | 3 | 510 | |||