Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GIT2 SIRNA Assays ›
Gene Symbol:
GIT2
Gene Name:
GIT ArfGAP 2
Gene Aliases:
CAT-2, CAT2, PKL
Chromosome Location:
Chr.12: 109929792 - 109996389 on Build GRCh38
UniGene ID:
Hs.434996
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
138620
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GIT2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135213.1
NP_001128685.1
3
419
NM_001135214.1
NP_001128686.1
3
419
NM_014776.3
NP_055591.2
3
419
NM_057169.3
NP_476510.1
3
419
NM_057170.3
NP_476511.1
3
419
NM_139201.2
NP_631940.2
3
419
XM_005253997.3
XP_005254054.1
3
414
XM_005254000.3
XP_005254057.1
3
414
XM_006719707.3
XP_006719770.1
3
414
XM_006719708.3
XP_006719771.1
3
414
XM_006719709.3
XP_006719772.1
3
414
XM_006719712.3
XP_006719775.1
3
414
XM_006719713.3
XP_006719776.1
3
411
XM_006719714.3
XP_006719777.1
3
414
XM_017020258.1
XP_016875747.1
3
414
XM_017020259.1
XP_016875748.1
3
411
XM_017020260.1
XP_016875749.1
3
411
XM_017020261.1
XP_016875750.1
3
409
GenBank mRNA
AB451294.1
-
3
254
AB451428.1
-
3
254
AF124491.1
-
3
254
AK222736.1
-
3
326
AK303198.1
-
2
120
AK304240.1
-
3
403
AK308304.1
-
3
334
BC001379.2
-
3
319
3051069
BC014223.2
-
2
170
BP871813.1
-
3
419
BT007312.1
-
3
254
CR533568.1
-
3
254
D63482.2
-
3
325
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of G-protein coupled receptor protein signaling pathway
positive regulation of GTPase activity
behavioral response to pain
GTPase activator activity
protein binding
protein complex binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0