Search
Search

TMEM204| Gene Symbol: TMEM204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256541.1 | NP_001243470.1 | 4 | 1302 | ||
| NM_024600.5 | NP_078876.2 | 3 | 1523 | |||
| GenBank mRNA | AK024551.1 | - | 3 | 1523 | ||
| AY676494.1 | - | 3 | 1524 | |||
| BC004932.1 | - | 4 | 1302 | 3622114 | ||
| BC105785.1 | - | 3 | 1023 | 4692469 | ||