Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other USP54 SIRNA Assays ›
Gene Symbol:
USP54
Gene Name:
ubiquitin specific peptidase 54
Gene Aliases:
C10orf29, bA137L10.3, bA137L10.4
Chromosome Location:
Chr.10: 73497538 - 73626844 on Build GRCh38
UniGene ID:
Hs.657355
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
148139
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: USP54
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001320437.1
NP_001307366.1
18
2928
NM_152586.3
NP_689799.3
18
3026
NR_135249.1
19
3413
NR_135250.1
14
2408
XM_005269582.2
XP_005269639.1
17
6104
XM_011539368.2
XP_011537670.1
18
6170
XM_017015759.1
XP_016871248.1
20
5599
XM_017015760.1
XP_016871249.1
20
4501
XM_017015761.1
XP_016871250.1
20
5339
XM_017015762.1
XP_016871251.1
20
4509
XM_017015763.1
XP_016871252.1
20
3803
XM_017015764.1
XP_016871253.1
20
3866
XM_017015765.1
XP_016871254.1
21
4119
XM_017015766.1
XP_016871255.1
20
5725
XM_017015767.1
XP_016871256.1
20
5384
XM_017015768.1
XP_016871257.1
20
3555
XM_017015769.1
XP_016871258.1
20
3163
XM_017015770.1
XP_016871259.1
20
3824
XM_017015771.1
XP_016871260.1
19
6341
XM_017015772.1
XP_016871261.1
19
6347
XM_017015773.1
XP_016871262.1
18
6281
XM_017015774.1
XP_016871263.1
19
5267
XM_017015775.1
XP_016871264.1
20
5145
XM_017015776.1
XP_016871265.1
19
5650
XM_017015777.1
XP_016871266.1
19
5309
XM_017015778.1
XP_016871267.1
18
6227
XM_017015779.1
XP_016871268.1
19
6347
XM_017015780.1
XP_016871269.1
19
6347
XM_017015781.1
XP_016871270.1
18
6176
XM_017015782.1
XP_016871271.1
19
3927
XM_017015783.1
XP_016871272.1
19
4974
XM_017015784.1
XP_016871273.1
18
2920
XM_017015785.1
XP_016871274.1
18
6227
XM_017015786.1
XP_016871275.1
18
3417
GenBank mRNA
AK094637.1
-
1
120
AK126497.1
-
6
1381
AK127213.1
-
5
961
AK127267.1
-
14
2408
AK303633.1
-
3
619
AL832102.1
-
1
66
BC064967.1
-
1
62
BC130633.1
-
3
610
Back To Top
More Information
Panther Classification:
Molecular Function -
cysteine protease
Gene Ontology Categories:
Function(s)
Process(es)
protein deubiquitination
protein binding
thiol-dependent ubiquitinyl hydrolase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0