Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ABI2 SIRNA Assays ›
          Gene Symbol:
          ABI2
          Gene Name:
          abl-interactor 2
          Gene Aliases:
          ABI-2, ABI2B, AIP-1, AblBP3, SSH3BP2, argBP1, argBPIA, argBPIB
          Chromosome Location:
          Chr.2: 203327874 - 203432173 on Build GRCh38
          UniGene ID:
          Hs.471156
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 148368

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ABI2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282925.1 NP_001269854.1 3 678
          NM_001282926.1 NP_001269855.1 3 678
          NM_001282927.1 NP_001269856.1 3 354
          NM_001282932.1 NP_001269861.1 4 875
          NM_005759.5 NP_005750.4 3 678
          XM_005246217.2 XP_005246274.1 3 1043
          XM_005246218.2 XP_005246275.1 3 1043
          XM_005246221.2 XP_005246278.1 3 1043
          XM_005246225.2 XP_005246282.1 3 1043
          XM_005246226.2 XP_005246283.1 3 1043
          XM_006712174.2 XP_006712237.1 3 1043
          XM_006712177.2 XP_006712240.1 4 1119
          XM_006712179.2 XP_006712242.1 3 1043
          XM_006712180.2 XP_006712243.1 3 1043
          XM_006712181.2 XP_006712244.1 3 1043
          XM_006712182.2 XP_006712245.1 3 1043
          XM_006712185.1 XP_006712248.1 4 1119
          XM_006712186.2 XP_006712249.1 3 1043
          XM_011510472.2 XP_011508774.1 3 1043
          XM_011510473.2 XP_011508775.1 2 285
          XM_017003118.1 XP_016858607.1 3 1043
          XM_017003119.1 XP_016858608.1 3 1043
          XM_017003120.1 XP_016858609.1 3 1043
          XM_017003121.1 XP_016858610.1 3 1043
          XM_017003122.1 XP_016858611.1 3 1043
          XM_017003123.1 XP_016858612.1 3 1043
          XM_017003124.1 XP_016858613.1 3 1043
          XM_017003125.1 XP_016858614.1 3 1043
          XM_017003126.1 XP_016858615.1 3 1043
          XM_017003127.1 XP_016858616.1 3 1043
          XM_017003128.1 XP_016858617.1 4 1119
          XM_017003129.1 XP_016858618.1 3 1043
          XM_017003130.1 XP_016858619.1 3 1043
          XM_017003131.1 XP_016858620.1 3 1043
          XM_017003132.1 XP_016858621.1 3 1043
          XM_017003133.1 XP_016858622.1 4 1119
          XM_017003134.1 XP_016858623.1 3 1043
          XM_017003135.1 XP_016858624.1 3 1043
          XM_017003136.1 XP_016858625.1 3 1043
          XM_017003137.1 XP_016858626.1 4 1119
          XM_017003138.1 XP_016858627.1 3 1043
          XM_017003139.1 XP_016858628.1 4 1119
          XM_017003140.1 XP_016858629.1 4 1119
          XM_017003141.1 XP_016858630.1 3 1043
          XM_017003142.1 XP_016858631.1 3 1043
          XM_017003143.1 XP_016858632.1 4 1119
          XM_017003144.1 XP_016858633.1 3 1043
          XM_017003145.1 XP_016858634.1 3 1043
          XM_017003146.1 XP_016858635.1 3 1043
          GenBank mRNA AF260261.1 - 3 402
          AK299824.1 - 3 522
          AK302853.1 - 4 801
          AK316000.1 - 5 617
          BC001439.2 - 3 427 3138341
          BT009920.1 - 3 402
          U23435.1 - 3 315
          X95632.1 - 3 502

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          movement of cell or subcellular component
          cytoskeleton organization
          actin polymerization or depolymerization
          viral process
          cell migration
          Rac protein signal transduction
          peptidyl-tyrosine phosphorylation
          positive regulation of Arp2/3 complex-mediated actin nucleation
          DNA binding
          protein binding
          cytoskeletal adaptor activity
          SH3 domain binding
          kinase binding
          ubiquitin protein ligase binding
          protein complex binding
          Rac GTPase binding
          proline-rich region binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-p786m:80/100.66.72.7:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0