Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FCHO1 SIRNA Assays ›
          Gene Symbol:
          FCHO1
          Gene Name:
          FCH domain only 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 17747718 - 17788568 on Build GRCh38
          UniGene ID:
          Hs.96485
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 148527

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FCHO1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001161357.1 NP_001154829.1 8, 9 776
          NM_001161358.1 NP_001154830.1 7, 8 686
          NM_001161359.1 NP_001154831.1 6, 7 616
          NM_015122.2 NP_055937.1 8, 9 780
          XM_005259831.3 XP_005259888.1 7, 8 861
          XM_006722701.3 XP_006722764.1 7, 8 784
          XM_006722702.3 XP_006722765.1 7, 8 861
          XM_011527824.2 XP_011526126.1 8, 9 776
          XM_011527825.2 XP_011526127.1 8, 9 765
          XM_011527826.2 XP_011526128.1 8, 9 748
          XM_011527827.2 XP_011526129.1 6, 7 583
          XM_011527828.2 XP_011526130.1 7, 8 679
          XM_017026519.1 XP_016882008.1 8, 9 846
          XM_017026520.1 XP_016882009.1 8, 9 829
          XM_017026521.1 XP_016882010.1 7, 8 780
          XM_017026522.1 XP_016882011.1 8, 9 776
          XM_017026523.1 XP_016882012.1 8, 9 846
          XM_017026524.1 XP_016882013.1 7, 8 861
          XM_017026525.1 XP_016882014.1 7, 8 659
          XM_017026526.1 XP_016882015.1 8, 9 776
          XM_017026527.1 XP_016882016.1 7, 8 784
          XM_017026528.1 XP_016882017.1 8, 9 846
          GenBank mRNA AB006628.1 - 5, 6 550
          AK291410.1 - 7, 8 642
          AK303623.1 - 4, 5 478
          AK309403.1 - 4, 5 529
          AK316458.1 - 6, 7 587
          BC028021.1 - 8, 9 721 5218319
          BC041130.1 - 8, 9 730 5757146

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          clathrin coat assembly
          clathrin-mediated endocytosis
          protein binding
          AP-2 adaptor complex binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0