Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rab5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK089199.1 | - | 6 | 1297 | ||
AK134691.1 | - | 6 | 1183 | |||
AK159832.1 | - | 6 | 1232 | |||
AK167599.1 | - | 6 | 1232 | |||
AK170540.1 | - | 6 | 1252 | |||
AK172328.1 | - | 6 | 1189 | |||
BC023027.1 | - | 6 | 1198 | 5369566 | ||
BC027378.1 | - | 5 | 1004 | |||
BC029678.1 | - | 6 | 1182 | 5368061 |