Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SLIT2 SIRNA Assays ›
          Gene Symbol:
          Slit2
          Gene Name:
          slit homolog 2 (Drosophila)
          Gene Aliases:
          Drad-1, E030015M03Rik, E130320P19Rik, Slil3, mKIAA4141, slit-2
          Chromosome Location:
          Chr.5: 47983108 - 48306282 on Build GRCm38
          UniGene ID:
          Mm.289739
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 152410

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Slit2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291227.1 NP_001278156.1 10, 11 2880
          NM_001291228.1 NP_001278157.1 10, 11 2880
          NM_178804.4 NP_848919.3 10, 11 2868
          GenBank mRNA AF144628.1 - 10, 11 1229
          AK033292.1 - 3, 4 334
          AK144573.1 - 10, 11 2869
          AK220505.1 - 10, 11 1464
          BC079911.1 - 10, 11 993
          BC145487.1 - 10, 11 1224 9054050
          BC145488.1 - 10, 11 1224 9054051
          BC150779.1 - 10, 11 1224 9087690

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          immunoglobulin superfamily cell adhesion molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          metanephros development
          ureteric bud development
          in utero embryonic development
          negative regulation of protein phosphorylation
          cell migration involved in sprouting angiogenesis
          negative regulation of leukocyte chemotaxis
          chemotaxis
          multicellular organism development
          nervous system development
          axonogenesis
          axon guidance
          motor neuron axon guidance
          negative regulation of cell proliferation
          negative regulation of lamellipodium assembly
          negative regulation of endothelial cell migration
          negative regulation of gene expression
          negative regulation of smooth muscle cell migration
          single organismal cell-cell adhesion
          olfactory bulb development
          chemorepulsion involved in postnatal olfactory bulb interneuron migration
          corticospinal neuron axon guidance through spinal cord
          telencephalon cell migration
          cell differentiation
          negative regulation of cell growth
          negative regulation of cell migration
          negative regulation of axon extension
          negative regulation of actin filament polymerization
          retinal ganglion cell axon guidance
          dorsal/ventral axon guidance
          negative regulation of GTPase activity
          Roundabout signaling pathway
          positive regulation of apoptotic process
          negative regulation of vascular permeability
          branching morphogenesis of an epithelial tube
          neuron projection morphogenesis
          axon extension involved in axon guidance
          negative regulation of inflammatory response
          negative chemotaxis
          induction of negative chemotaxis
          negative regulation of small GTPase mediated signal transduction
          mammary gland duct morphogenesis
          mammary duct terminal end bud growth
          negative regulation of chemokine-mediated signaling pathway
          cellular response to heparin
          negative regulation of smooth muscle cell chemotaxis
          negative regulation of mononuclear cell migration
          negative regulation of neutrophil chemotaxis
          negative regulation of monocyte chemotaxis
          negative regulation of retinal ganglion cell axon guidance
          negative regulation of cellular response to growth factor stimulus
          GTPase inhibitor activity
          receptor binding
          calcium ion binding
          protein binding
          heparin binding
          identical protein binding
          protein homodimerization activity
          laminin-1 binding
          proteoglycan binding
          heparan sulfate proteoglycan binding
          chemorepellent activity
          Roundabout binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-wf5dr:80/100.66.77.5:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0