Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PDE1A SIRNA Assays ›
Gene Symbol:
PDE1A
Gene Name:
phosphodiesterase 1A
Gene Aliases:
CAM-PDE-1A, HCAM-1, HCAM1, HSPDE1A
Chromosome Location:
Chr.2: 182140035 - 182523375 on Build GRCh38
UniGene ID:
Hs.191046
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
15619
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PDE1A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001003683.2
NP_001003683.1
5, 6
664
NM_001258312.1
NP_001245241.1
5, 6
691
NM_001258313.1
NP_001245242.1
4, 5
843
NM_001258314.1
NP_001245243.1
3, 4
520
NM_005019.4
NP_005010.2
4, 5
706
XM_011511323.2
XP_011509625.1
4, 5
483
XM_011511324.2
XP_011509626.1
5, 6
711
XM_011511325.2
XP_011509627.1
4, 5
838
XM_011511326.2
XP_011509628.1
3, 4
515
XM_017004294.1
XP_016859783.1
4, 5
706
XM_017004295.1
XP_016859784.1
5, 6
711
XM_017004296.1
XP_016859785.1
5, 6
633
XM_017004297.1
XP_016859786.1
5, 6
574
XM_017004298.1
XP_016859787.1
5, 6
756
XM_017004299.1
XP_016859788.1
5, 6
711
XM_017004300.1
XP_016859789.1
5, 6
756
XM_017004301.1
XP_016859790.1
5, 6
561
XM_017004302.1
XP_016859791.1
4, 5
838
GenBank mRNA
AJ401610.1
-
4, 5
425
AK294239.1
-
5, 6
626
AK295657.1
-
5, 6
496
AK301720.1
-
3, 4
503
AL110263.1
-
4, 5
817
BC022480.1
-
4, 5
674
4814064
DC396228.1
-
3, 4
520
U40370.1
-
5, 6
557
Back To Top
More Information
Panther Classification:
Molecular Function -
phosphodiesterase
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
3',5'-cyclic-AMP phosphodiesterase activity
calmodulin-dependent cyclic-nucleotide phosphodiesterase activity
calmodulin binding
metal ion binding
3',5'-cyclic-GMP phosphodiesterase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline