Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ovol1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019935.3 | NP_064319.1 | 4 | 2512 | ||
GenBank mRNA | AK028577.1 | - | 4 | 2513 | ||
AK132505.1 | - | 4 | 2552 |