Search
Search

F8| Gene Symbol: F8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161373.1 | NP_001154845.1 | 6, 7 | 1207 | ||
| NM_001161374.1 | NP_001154846.1 | 6, 7 | 1207 | |||
| NM_007977.2 | NP_032003.2 | 6, 7 | 1207 | |||
| GenBank mRNA | AJ627040.1 | - | 5, 6 | 627 | ||
| AK051579.1 | - | 6, 7 | 3483 | |||
| BC138347.1 | - | 6, 7 | 1088 | 8861367 | ||
| BC145245.1 | - | 6, 7 | 1088 | 9053797 | ||
| L05573.1 | - | 6, 7 | 1205 | |||