Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ST5 SIRNA Assays ›
Gene Symbol:
ST5
Gene Name:
suppression of tumorigenicity 5
Gene Aliases:
DENND2B, HTS1, p126
Chromosome Location:
Chr.11: 8693352 - 8910951 on Build GRCh38
UniGene ID:
Hs.117715
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
16243
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ST5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_005418.3
NP_005409.3
23
4245
NM_139157.2
NP_631896.1
19
2713
NM_213618.1
NP_998783.1
20
4045
XM_005253077.1
XP_005253134.1
20
3960
XM_005253083.2
XP_005253140.1
19
2855
XM_011520309.1
XP_011518611.1
21
4020
XM_011520310.1
XP_011518612.1
21
4432
XM_011520311.2
XP_011518613.1
21
4414
XM_011520312.1
XP_011518614.1
25
4379
XM_011520313.1
XP_011518615.1
24
4265
XM_011520314.1
XP_011518616.1
23
4157
XM_011520315.1
XP_011518617.1
23
4253
XM_011520316.1
XP_011518618.1
22
4183
XM_011520317.1
XP_011518619.1
21
4124
XM_011520318.1
XP_011518620.1
24
4319
XM_011520319.2
XP_011518621.1
21
3972
XM_011520323.2
XP_011518625.2
20
2843
XM_011520324.2
XP_011518626.2
19
2783
XM_011520325.1
XP_011518627.1
20
2915
XM_011520326.1
XP_011518628.1
24
3119
XM_011520327.1
XP_011518629.1
20
2808
XM_011520328.1
XP_011518630.1
20
2840
XM_011520329.1
XP_011518631.1
23
3059
XM_017018182.1
XP_016873671.1
22
4138
XM_017018183.1
XP_016873672.1
22
4115
XM_017018184.1
XP_016873673.1
24
4274
XM_017018185.1
XP_016873674.1
20
4070
XM_017018186.1
XP_016873675.1
19
4010
XM_017018187.1
XP_016873676.1
20
2779
GenBank mRNA
AK126554.1
-
8
1957
AK127763.1
-
17
3100
BC036655.1
-
23
4254
4837231
BX647431.1
-
20
4045
S45936.1
-
16
2387
U15131.1
-
20
3977
U15779.1
-
16
2346
U15780.1
-
19
2717
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of GTPase activity
Rab guanyl-nucleotide exchange factor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0