Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Vat1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173016.3 | NP_766604.2 | 1, 2 | 307 | ||
GenBank mRNA | AK122527.1 | - | 1, 2 | 307 | ||
BC056927.1 | - | 1, 2 | 317 | 5719211 |