Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Dnajc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253685.1 | NP_001240614.1 | 2 | 141 | ||
NM_013888.3 | NP_038916.1 | 2 | 278 | |||
GenBank mRNA | AF132906.2 | - | 2 | 227 | ||
AK032207.1 | - | 2 | 272 | |||
BC013501.1 | - | 2 | 185 | 4222828 | ||
BC071211.1 | - | 2 | 166 | |||
BF453706.1 | - | 2 | 141 | |||
DV042983.1 | - | 2 | 293 |