Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ALDOB SIRNA Assays ›
          Gene Symbol:
          Aldob
          Gene Name:
          aldolase B, fructose-bisphosphate
          Gene Aliases:
          Aldo-2, Aldo2, BC016435
          Chromosome Location:
          Chr.4: 49535995 - 49549483 on Build GRCm38
          UniGene ID:
          Mm.482116
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 169565

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Aldob
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_144903.2 NP_659152.1 9 1230
          GenBank mRNA AK135385.1 - 9 1259
          AK149395.1 - 9 1247
          AK149598.1 - 9 1244
          AK167334.1 - 9 1247
          AK167566.1 - 9 1244
          AK168600.1 - 9 1244
          AK168997.1 - 9 1294
          BC016435.1 - 9 1226 4241025
          BC022113.1 - 9 1203 5098524
          BC024056.1 - 9 1236 5100067
          BC024112.1 - 9 1235 5102118
          BC026577.1 - 9 1195 4989521
          BC030724.1 - 9 1232 5100784
          BC030725.1 - 9 1236 5101686
          BC034169.1 - 9 1236 4975382
          BC034171.1 - 9 1213 4972521
          BC034172.1 - 9 1200 4974489
          BC034173.1 - 9 1212 4974553
          BC036130.1 - 9 1213 4974071
          BC036131.1 - 9 1230 4974877
          BC036132.1 - 9 1230 4975336
          BC036133.1 - 9 1242 4975573
          BC036139.1 - 5 854
          BC036142.1 - 3 537

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          aldolase

          Gene Ontology Categories:

          Function(s) Process(es)

          fructose metabolic process
          glycolytic process
          NADH oxidation
          response to zinc ion
          fructose 1,6-bisphosphate metabolic process
          positive regulation of ATPase activity
          response to peptide hormone
          vacuolar proton-transporting V-type ATPase complex assembly
          catalytic activity
          fructose-bisphosphate aldolase activity
          cytoskeletal protein binding
          lyase activity
          phosphatidylcholine binding
          identical protein binding
          ATPase binding
          fructose-1-phosphate aldolase activity
          fructose binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0