Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ERN1 SIRNA Assays ›
Gene Symbol:
Ern1
Gene Name:
endoplasmic reticulum (ER) to nucleus signalling 1
Gene Aliases:
9030414B18Rik, AI225830, C85377, Ire1a, Ire1alpha, Ire1p
Chromosome Location:
Chr.11: 106397620 - 106487796 on Build GRCm38
UniGene ID:
Mm.340943
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
174323
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ern1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_023913.2
NP_076402.1
14
1874
GenBank mRNA
AB031332.1
-
14
1874
AK077041.1
-
1896
AK080709.1
-
8
1127
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001191926, XM_006247634, XM_006247633.
Panther Classification:
Molecular Function -
endoribonuclease
Gene Ontology Categories:
Function(s)
Process(es)
endothelial cell proliferation
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA cleavage
mRNA processing
protein phosphorylation
apoptotic process
response to unfolded protein
activation of signaling protein activity involved in unfolded protein response
cell cycle arrest
activation of JUN kinase activity
metabolic process
phosphorylation
endoplasmic reticulum unfolded protein response
mRNA splicing via endonucleolytic cleavage and ligation involved in unfolded protein response
positive regulation of RNA splicing
response to endoplasmic reticulum stress
cellular response to vascular endothelial growth factor stimulus
peptidyl-serine autophosphorylation
IRE1-mediated unfolded protein response
protein autophosphorylation
mRNA splicing, via endonucleolytic cleavage and ligation
mRNA endonucleolytic cleavage involved in unfolded protein response
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
cellular response to glucose stimulus
RNA phosphodiester bond hydrolysis, endonucleolytic
positive regulation of endoplasmic reticulum unfolded protein response
insulin metabolic process
peptidyl-serine trans-autophosphorylation
nucleotide binding
magnesium ion binding
catalytic activity
endonuclease activity
endoribonuclease activity
ribonuclease activity
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
kinase activity
bidentate ribonuclease III activity
transferase activity
transferase activity, transferring phosphorus-containing groups
hydrolase activity
enzyme binding
Hsp70 protein binding
identical protein binding
protein homodimerization activity
ADP binding
metal ion binding
Hsp90 protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-9cgzg:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0