Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cttnbp2nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163332.1 | NP_001156804.1 | 6 | 2422 | ||
NM_001163333.1 | NP_001156805.1 | 6 | 2414 | |||
NM_030249.4 | NP_084525.1 | 6 | 2530 | |||
GenBank mRNA | AK029901.1 | - | 6 | 2532 | ||
AK030438.1 | - | 6 | 2395 | |||
AK129359.1 | - | 6 | 2420 | |||
AK145085.1 | - | 6 | 2396 | |||
BC003236.1 | - | 6 | 2384 | 3498078 | ||
BC006952.1 | - | 2 | 1872 |