Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc19a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030556.2 | NP_085033.2 | 5 | 1346 | ||
GenBank mRNA | AF271634.1 | - | 5 | 1346 | ||
AK079566.1 | - | 2 | 265 | |||
AK098112.1 | - | 5 | 1337 | |||
BC109154.1 | - | 5 | 1329 | 40047017 | ||
BC109155.1 | - | 4 | 1199 | 40047019 |