Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Plekha3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031256.3 | NP_112546.1 | 5 | 835 | ||
GenBank mRNA | AF286163.1 | - | 5 | 624 | ||
BC012283.1 | - | 4 | 1094 | |||
BC031110.1 | - | 6 | 643 | 4981486 |