Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tmem2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033759.2 | NP_001028931.1 | 19 | 3743 | ||
NM_031997.4 | NP_114386.3 | 20 | 3825 | |||
GenBank mRNA | AF137031.1 | - | 2 | 188 | ||
AK129352.1 | - | 14 | 2044 | |||
BC019745.1 | - | 11 | 1466 | 3675225 | ||
BC076570.1 | - | 19 | 3349 | 6830791 | ||
BC089353.1 | - | 19 | 3725 | 6410437 | ||
BG244335.1 | - | 441 | ||||
BI666286.1 | - | 3 | 360 |