Search
Search

Shroom1| Gene Symbol: Shroom1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290789.1 | NP_001277718.1 | 7 | 2413 | ||
| NM_027917.3 | NP_082193.2 | 8 | 2602 | |||
| GenBank mRNA | AK004934.1 | - | 7 | 2641 | ||
| AK035462.1 | - | 3 | 3852 | |||
| AK085693.1 | - | 6 | 2457 | |||
| AK134332.1 | - | 8 | 2532 | |||
| BC005761.1 | - | 7 | 2388 | 3601742 | ||
| BC031598.1 | - | 7 | 2305 | 3497170 | ||