Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PDLIM5 SIRNA Assays ›
          Gene Symbol:
          PDLIM5
          Gene Name:
          PDZ and LIM domain 5
          Gene Aliases:
          ENH, ENH1, L9, LIM
          Chromosome Location:
          Chr.4: 94451857 - 94668227 on Build GRCh38
          UniGene ID:
          Hs.480311
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 17950

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PDLIM5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001011513.3 NP_001011513.3 2 255
          NM_001011515.2 NP_001011515.1 2 255
          NM_001011516.2 NP_001011516.1 2 255
          NM_001256425.1 NP_001243354.1 2 255
          NM_001256426.1 NP_001243355.1 2 255
          NM_001256427.1 NP_001243356.1 2 255
          NM_001256428.1 NP_001243357.1 2 255
          NM_001256429.1 NP_001243358.1 2 255
          NM_006457.4 NP_006448.4 2 255
          NR_046186.1 2 255
          XM_005262693.4 XP_005262750.1 2 225
          XM_005262695.4 XP_005262752.1 2 225
          XM_005262696.4 XP_005262753.1 2 170
          XM_005262698.4 XP_005262755.1 2 177
          XM_006714066.3 XP_006714129.1 2 225
          XM_006714068.3 XP_006714131.1 2 170
          XM_006714069.3 XP_006714132.1 2 225
          XM_006714070.3 XP_006714133.1 2 228
          XM_011531543.2 XP_011529845.1 2 225
          XM_017007657.1 XP_016863146.1 2 225
          XM_017007658.1 XP_016863147.1 2 170
          GenBank mRNA AF061258.1 - 2 157
          AK291624.1 - 2 177
          AK296980.1 - 2 169
          AK304125.1 - 2 177
          AL832157.1 - 2 157
          AL832686.1 - 2 159
          AL833151.1 - 2 153
          AL833438.1 - 2 140
          AY345240.1 - 2 227
          AY598328.1 - 1 74
          BC008741.2 - 2 194 3345715
          BC017902.1 - 2 198
          BC026029.2 - 1 117
          BF979604.1 - 2 225
          BP260089.1 - 2 212
          BT007328.1 - 1 74
          D31139.1 - 2 236
          DA381128.1 - 2 175
          DA895978.1 - 2 255

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_053326, XM_006233415, XM_006233404, XM_006233413, XM_006233419, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001190853, NM_001190855, NM_001190854, NM_001190857, NM_019809, and others.

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of synapse assembly
          regulation of dendritic spine morphogenesis
          cell growth involved in cardiac muscle cell development
          cell-cell adhesion
          actin binding
          protein kinase C binding
          protein binding
          zinc ion binding
          actinin binding
          protein N-terminus binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0