Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RUVBL2 SIRNA Assays ›
          Gene Symbol:
          RUVBL2
          Gene Name:
          RuvB like AAA ATPase 2
          Gene Aliases:
          CGI-46, ECP-51, ECP51, INO80J, REPTIN, RVB2, TAP54-beta, TIH2, TIP48, TIP49B
          Chromosome Location:
          Chr.19: 48993448 - 49015995 on Build GRCh38
          UniGene ID:
          Hs.515846
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 18429

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RUVBL2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001321190.1 NP_001308119.1 13 1670
          NM_001321191.1 NP_001308120.1 13 1579
          NM_006666.2 NP_006657.1 13 1600
          NR_135578.1 13 1589
          XM_011526330.1 XP_011524632.1 13 1513
          GenBank mRNA AB024301.1 - 13 1149
          AF124607.1 - 13 1157
          AF151804.1 - 13 1142
          AF155138.1 - 13 1142
          AK027762.1 - 13 1220
          AK057498.1 - 13 2147
          AK074542.1 - 13 1150
          AL136743.1 - 12 1139
          BC000428.2 - 13 1141 2819778
          BC004531.2 - 13 1149 3051517
          BC008355.1 - 13 1138
          CR533507.1 - 12 1136
          Y18417.1 - 13 1141

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          box C/D snoRNP assembly
          DNA repair
          DNA recombination
          chromatin remodeling
          transcription, DNA-templated
          protein folding
          DNA duplex unwinding
          cellular response to UV
          positive regulation of histone acetylation
          regulation of growth
          histone H4 acetylation
          histone H2A acetylation
          positive regulation of transcription from RNA polymerase II promoter
          establishment of protein localization to chromatin
          cellular response to estradiol stimulus
          transcriptional activation by promoter-enhancer looping
          negative regulation of estrogen receptor binding
          positive regulation of telomerase RNA localization to Cajal body
          RNA polymerase II core promoter sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          TFIID-class transcription factor binding
          DNA helicase activity
          ATP-dependent DNA helicase activity
          protein binding
          ATP binding
          ATPase activity
          chromatin DNA binding
          identical protein binding
          protein homodimerization activity
          ATP-dependent 5'-3' DNA helicase activity
          ADP binding
          unfolded protein binding
          ATPase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0