Search
Search

Cnot4| Gene Symbol: Cnot4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164411.1 | NP_001157883.1 | 5 | 850 | ||
| NM_001164412.1 | NP_001157884.1 | 5 | 850 | |||
| NM_001164413.1 | NP_001157885.1 | 5 | 850 | |||
| NM_001164414.1 | NP_001157886.1 | 5 | 850 | |||
| NM_016877.4 | NP_058573.3 | 5 | 850 | |||
| GenBank mRNA | AK029753.1 | - | 5 | 818 | ||
| AK032248.1 | - | 5 | 830 | |||
| AK075887.1 | - | 5 | 839 | |||
| AK144760.1 | - | 5 | 808 | |||
| AK165997.1 | - | 5 | 836 | |||
| BC048243.1 | - | 5 | 738 | 6493673 | ||
| BC058778.1 | - | 5 | 802 | 5397231 | ||
| U71269.1 | - | 5 | 836 | |||