Search
Search

Pmfbp1| Gene Symbol: Pmfbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019938.3 | NP_064322.1 | 3 | 192 | ||
| GenBank mRNA | AB029919.1 | - | 3 | 192 | ||
| AK133053.1 | - | 2 | 248 | |||
| BC138245.1 | - | 3 | 144 | 8861264 | ||
| BC138246.1 | - | 3 | 144 | 8861265 | ||