Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Azi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286508.1 | NP_001273437.1 | 8 | 2787 | ||
NM_013727.4 | NP_038755.1 | 8 | 2869 | |||
GenBank mRNA | AB007141.1 | - | 8 | 2704 | ||
AK030076.1 | - | 8 | 2701 | |||
AK039794.1 | - | 1 | 986 | |||
BC006851.1 | - | 8 | 2764 | 3156857 |