Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ADRB2 SIRNA Assays ›
          Gene Symbol:
          ADRB2
          Gene Name:
          adrenoceptor beta 2
          Gene Aliases:
          ADRB2R, ADRBR, B2AR, BAR, BETA2AR
          Chromosome Location:
          Chr.5: 148826593 - 148828634 on Build GRCh38
          UniGene ID:
          Hs.2551
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 1894

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ADRB2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000024.5 NP_000015.1 1 1458
          GenBank mRNA AK223025.1 - 1 1421
          AK313151.1 - 1 1440
          AY136741.1 - 1 1219
          BC012481.2 - 1 1428 4538187
          BC063486.1 - 1 1406 5241774
          BC073856.1 - 1 1409 5217922
          KJ534762.1 - 1 1219
          M15169.1 - 1 2806
          X04827.1 - 1 1396

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          diet induced thermogenesis
          vasodilation by norepinephrine-epinephrine involved in regulation of systemic arterial blood pressure
          desensitization of G-protein coupled receptor protein signaling pathway by arrestin
          diaphragm contraction
          positive regulation of the force of heart contraction by epinephrine
          receptor-mediated endocytosis
          cell surface receptor signaling pathway
          activation of transmembrane receptor protein tyrosine kinase activity
          adenylate cyclase-modulating G-protein coupled receptor signaling pathway
          activation of adenylate cyclase activity
          cell-cell signaling
          female pregnancy
          aging
          positive regulation of cell proliferation
          associative learning
          endosome to lysosome transport
          response to cold
          positive regulation of sodium ion transport
          negative regulation of angiogenesis
          negative regulation of ossification
          positive regulation of bone mineralization
          positive regulation of protein ubiquitination
          heat generation
          response to progesterone
          positive regulation of ATPase activity
          response to testosterone
          synaptic transmission, glutamatergic
          negative regulation of urine volume
          negative regulation of multicellular organism growth
          wound healing
          positive regulation of apoptotic process
          positive regulation of potassium ion transport
          positive regulation of MAPK cascade
          response to estrogen
          estrous cycle
          bone resorption
          positive regulation of vasodilation
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of smooth muscle contraction
          positive regulation of skeletal muscle tissue growth
          negative regulation of inflammatory response
          brown fat cell differentiation
          regulation of calcium ion transport
          regulation of sensory perception of pain
          excitatory postsynaptic potential
          cellular response to hypoxia
          response to dexamethasone
          response to monoamine
          adenylate cyclase-activating adrenergic receptor signaling pathway
          negative regulation of platelet aggregation
          liver regeneration
          positive regulation of autophagosome maturation
          positive regulation of lipophagy
          positive regulation of mitophagy in response to mitochondrial depolarization
          G-protein alpha-subunit binding
          beta2-adrenergic receptor activity
          protein binding
          drug binding
          adenylate cyclase binding
          potassium channel regulator activity
          B2 bradykinin receptor binding
          protein complex binding
          dopamine binding
          ionotropic glutamate receptor binding
          protein homodimerization activity
          epinephrine binding
          norepinephrine binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-mtn5c:80/100.66.73.196:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0