Search
Search

Ctnnb1| Gene Symbol: Ctnnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053357.2 | NP_445809.2 | 3 | 479 | ||
| XM_008766691.1 | XP_008764913.1 | 3 | 562 | |||
| GenBank mRNA | AF121265.1 | - | 3 | 479 | ||
| FQ222424.1 | - | 672 | ||||
| FQ224468.1 | - | 677 | ||||