Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: OIP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007280.1 | NP_009211.1 | 2, 3 | 453 | ||
GenBank mRNA | AF025441.1 | - | 2, 3 | 399 | ||
BC015050.1 | - | 2, 3 | 453 | 3908972 |