Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other AHSG SIRNA Assays ›
Gene Symbol:
Ahsg
Gene Name:
alpha-2-HS-glycoprotein
Gene Aliases:
Aa2-066, pp63
Chromosome Location:
Chr.11: 81711269 - 81717594 on Build Rnor_6.0
UniGene ID:
Rn.32083
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
197251
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ahsg
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012898.4
NP_037030.1
4
614
GenBank mRNA
AY325170.1
-
1250
BC091118.1
-
4
592
7368203
FQ209390.1
-
4
611
FQ209416.1
-
4
613
FQ209417.1
-
4
614
FQ209422.1
-
4
613
FQ209445.1
-
4
613
FQ209451.1
-
4
613
FQ209502.1
-
4
613
FQ209536.1
-
4
613
FQ209545.1
-
4
613
FQ209557.1
-
4
613
FQ209568.1
-
4
613
FQ209581.1
-
4
613
FQ209585.1
-
4
613
FQ209588.1
-
4
613
FQ209604.1
-
4
613
FQ209612.1
-
4
613
FQ209617.1
-
4
613
FQ209654.1
-
4
613
FQ209655.1
-
4
610
FQ209665.1
-
4
613
FQ209680.1
-
4
613
FQ209720.1
-
4
613
FQ209759.1
-
4
613
FQ209767.1
-
4
622
FQ209793.1
-
4
613
FQ209828.1
-
4
568
FQ209858.1
-
4
611
FQ209920.1
-
4
613
FQ209985.1
-
4
613
FQ210030.1
-
4
608
FQ210033.1
-
4
613
FQ210056.1
-
4
611
FQ210082.1
-
4
613
FQ210085.1
-
4
611
FQ210120.1
-
4
612
FQ210139.1
-
4
613
FQ210141.1
-
4
613
FQ210159.1
-
4
613
FQ210164.1
-
4
611
FQ210170.1
-
4
613
FQ210199.1
-
4
613
FQ210228.1
-
4
613
FQ210239.1
-
4
613
FQ210242.1
-
4
610
FQ210249.1
-
4
613
FQ210260.1
-
4
613
FQ210304.1
-
4
613
FQ210382.1
-
4
613
FQ210396.1
-
4
613
FQ210402.1
-
4
613
FQ210405.1
-
4
613
FQ210452.1
-
4
614
FQ210457.1
-
4
613
FQ210498.1
-
4
613
FQ210525.1
-
4
612
FQ210533.1
-
2
215
FQ210562.1
-
4
613
FQ210566.1
-
4
613
FQ210623.1
-
4
613
FQ210700.1
-
4
613
FQ210709.1
-
4
612
FQ210731.1
-
4
611
FQ210738.1
-
4
613
FQ210739.1
-
4
613
FQ210760.1
-
4
611
FQ210764.1
-
4
613
FQ210802.1
-
3
276
FQ210871.1
-
4
613
FQ210891.1
-
4
613
FQ210896.1
-
4
611
FQ210953.1
-
4
613
FQ210957.1
-
4
619
FQ210983.1
-
4
613
FQ211020.1
-
4
613
FQ211045.1
-
4
610
FQ211090.1
-
4
613
FQ211092.1
-
4
613
FQ211115.1
-
4
613
FQ211177.1
-
4
613
FQ211179.1
-
4
613
FQ211283.1
-
4
613
FQ211299.1
-
4
613
FQ211301.1
-
4
614
FQ211303.1
-
4
613
FQ211304.1
-
4
613
FQ211368.1
-
4
613
FQ211379.1
-
4
613
FQ211387.1
-
403
FQ211843.1
-
4
613
FQ217075.1
-
4
611
FQ218089.1
-
4
613
FQ218223.1
-
4
612
FQ218236.1
-
4
613
FQ218262.1
-
4
613
FQ218264.1
-
4
613
FQ218269.1
-
4
613
FQ218319.1
-
2
212
FQ218332.1
-
4
613
FQ218341.1
-
4
613
FQ218342.1
-
4
611
FQ218355.1
-
4
612
FQ218373.1
-
4
613
FQ218390.1
-
4
613
FQ218400.1
-
4
613
FQ218410.1
-
420
FQ218424.1
-
4
611
FQ218436.1
-
4
611
FQ218463.1
-
4
611
FQ218464.1
-
4
613
FQ218469.1
-
4
611
FQ218479.1
-
4
613
FQ218480.1
-
4
613
FQ218487.1
-
4
613
FQ218489.1
-
4
613
FQ218493.1
-
4
613
FQ218500.1
-
4
613
FQ218502.1
-
4
613
FQ218509.1
-
4
611
FQ218531.1
-
4
613
FQ218533.1
-
4
613
FQ218563.1
-
4
611
FQ218569.1
-
4
611
FQ218570.1
-
4
613
FQ218580.1
-
4
613
FQ218585.1
-
4
613
FQ218608.1
-
4
613
FQ218609.1
-
4
613
FQ218630.1
-
4
609
FQ218634.1
-
4
613
FQ218648.1
-
4
611
FQ218687.1
-
4
441
FQ218695.1
-
4
613
FQ218713.1
-
4
611
FQ218744.1
-
4
595
FQ218765.1
-
4
613
FQ218787.1
-
4
615
FQ218796.1
-
4
613
FQ218800.1
-
4
613
FQ218813.1
-
4
700
FQ218846.1
-
4
613
FQ218858.1
-
4
610
FQ218862.1
-
4
613
FQ218872.1
-
4
613
FQ218877.1
-
4
611
FQ218889.1
-
4
613
FQ218890.1
-
4
613
FQ218892.1
-
4
613
FQ218907.1
-
4
613
FQ218912.1
-
4
612
FQ218931.1
-
4
612
FQ218948.1
-
4
613
FQ218951.1
-
4
613
FQ218970.1
-
4
611
FQ218974.1
-
4
614
FQ218975.1
-
4
613
FQ218980.1
-
4
613
FQ218981.1
-
4
611
FQ219018.1
-
4
613
FQ219028.1
-
4
613
FQ219033.1
-
4
613
FQ219042.1
-
4
612
FQ219060.1
-
4
613
FQ219061.1
-
4
613
FQ219086.1
-
4
613
FQ219094.1
-
4
613
FQ219099.1
-
4
613
FQ219103.1
-
4
613
FQ219110.1
-
4
613
FQ219121.1
-
4
613
FQ219129.1
-
4
611
FQ219130.1
-
4
609
FQ219141.1
-
4
613
FQ219164.1
-
4
613
FQ219173.1
-
4
613
FQ219194.1
-
4
613
FQ219203.1
-
4
613
FQ219221.1
-
4
613
FQ219232.1
-
4
619
FQ219235.1
-
4
613
FQ219239.1
-
4
613
FQ219251.1
-
4
613
FQ219274.1
-
4
613
FQ219295.1
-
4
613
FQ219300.1
-
4
613
FQ219311.1
-
4
613
FQ219320.1
-
4
613
FQ219326.1
-
4
613
FQ219358.1
-
4
612
FQ219363.1
-
4
615
FQ219364.1
-
4
613
FQ219369.1
-
4
611
FQ219370.1
-
4
613
FQ219377.1
-
4
613
FQ219388.1
-
4
613
FQ219400.1
-
4
613
FQ219406.1
-
4
613
FQ219412.1
-
4
613
FQ219419.1
-
4
611
FQ219486.1
-
4
608
FQ219487.1
-
4
613
FQ219488.1
-
4
613
FQ219498.1
-
4
611
FQ219536.1
-
4
612
FQ219546.1
-
4
613
FQ219551.1
-
4
613
FQ219552.1
-
4
611
FQ219558.1
-
4
613
FQ219565.1
-
4
613
FQ219587.1
-
4
611
FQ219606.1
-
4
613
FQ219607.1
-
4
610
FQ219608.1
-
4
611
FQ219616.1
-
4
348
FQ219617.1
-
4
613
FQ219643.1
-
4
613
FQ219653.1
-
4
612
FQ219660.1
-
4
613
FQ219663.1
-
4
613
FQ219668.1
-
4
614
FQ219672.1
-
4
613
FQ219688.1
-
4
611
FQ219691.1
-
4
613
FQ219726.1
-
4
613
FQ219755.1
-
4
613
FQ219769.1
-
4
613
FQ219770.1
-
4
613
FQ219772.1
-
4
612
FQ219773.1
-
4
613
FQ219774.1
-
4
613
FQ222367.1
-
4
611
M29758.1
-
4
614
X63446.1
-
4
572
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001276450, NM_013465, AK133523, BC012678, BC019822, and others.
Panther Classification:
Molecular Function -
protease inhibitor
Gene Ontology Categories:
Function(s)
Process(es)
ossification
protein complex assembly
acute-phase response
male gonad development
negative regulation of endopeptidase activity
cerebral cortex development
negative regulation of cell growth
negative regulation of bone mineralization
organ regeneration
cellular response to insulin stimulus
negative regulation of phosphorylation
positive regulation of bone resorption
negative regulation of insulin receptor signaling pathway
regulation of inflammatory response
positive regulation of phagocytosis
negative regulation of protein tyrosine kinase activity
cysteine-type endopeptidase inhibitor activity
kinase inhibitor activity
receptor signaling protein tyrosine kinase inhibitor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline